Assay Details
Gene Symbol: Rpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100794.1 | NP_001094264.1 | 8 - 9 | 1038 | 121 | |
GenBank mRNA | BC166888.1 | - | 8 - 9 | 1035 | 121 | |
BC166920.1 | - | 8 - 9 | 1035 | 121 | ||
BI299235.1 | - | 1 - 2 | 121 | |||
FQ212650.1 | - | 8 - 9 | 1017 | 121 | ||
FQ226064.1 | - | 8 - 9 | 1024 | 121 | ||
FQ234387.1 | - | 8 - 9 | 1024 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
