Assay Details
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127544.1 | NP_001121016.1 | 1 - 2 | 245 | 120 | |
XM_008761404.1 | XP_008759626.1 | 1 - 2 | 361 | 120 | ||
GenBank mRNA | BC162022.1 | - | 1 - 2 | 224 | 120 | |
FQ211892.1 | - | 1 - 2 | 293 | 120 | ||
FQ221591.1 | - | 1 - 2 | 325 | 120 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008761404.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
