Assay Details
Gene Symbol: Metap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106476.1 | NP_001099946.1 | 5 - 6 | 514 | 106 | |
GenBank mRNA | FQ225038.1 | - | 5 - 6 | 437 | 106 | |
FQ227458.1 | - | 5 - 6 | 437 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
