Assay Details
Gene Symbol: Ppa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135871.1 | NP_001129343.1 | 3 - 4 | 244 | 80 | |
XM_006233327.2 | XP_006233389.1 | 3 - 4 | 1841 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
