Assay Details
Gene Symbol: Znhit6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106203.2 | NP_001099673.1 | 3 - 4 | 837 | 61 | |
GenBank mRNA | BC161957.1 | - | 3 - 4 | 837 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
