Assay Details
Gene Symbol: Dennd2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107714.1 | NP_001101184.1 | 10 - 11 | 1176 | 82 | |
XM_006233114.2 | XP_006233176.1 | 10 - 11 | 1125 | 82 | ||
XM_008761396.1 | XP_008759618.1 | 9 - 10 | 1572 | 82 | ||
XM_008761397.1 | XP_008759619.1 | 9 - 10 | 1572 | 82 | ||
XM_008761398.1 | XP_008759620.1 | 10 - 11 | 1125 | 82 | ||
XM_008761399.1 | XP_008759621.1 | 10 - 11 | 1209 | 82 | ||
GenBank mRNA | BC158880.1 | - | 10 - 11 | 1112 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
