Assay Details
Gene Symbol: Dennd2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107714.1 | NP_001101184.1 | 11 - 12 | 1413 | 125 | |
XM_006233114.2 | XP_006233176.1 | 11 - 12 | 1362 | 125 | ||
XM_008761397.1 | XP_008759619.1 | 10 - 11 | 1809 | 125 | ||
GenBank mRNA | BC158880.1 | - | 11 - 12 | 1349 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
