Assay Details
Gene Symbol: Prss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053504.1 | NP_445956.1 | 7 - 8 | 1675 | 100 | |
GenBank mRNA | AJ311671.2 | - | 7 - 8 | 1675 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
