Assay Details
Gene Symbol: Kat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750617.3 | XP_003750665.2 | 14 - 15 | 2924 | 57 | |
GenBank mRNA | BC092639.1 | - | 1 - 2 | 329 | 57 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC092639.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
