Assay Details
Gene Symbol: Agpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133406.1 | NP_596897.1 | 4 - 5 | 711 | 85 | |
XM_006227856.2 | XP_006227918.1 | 4 - 5 | 1426 | 85 | ||
XM_006227857.2 | XP_006227919.1 | 4 - 5 | 1080 | 85 | ||
GenBank mRNA | AB067572.1 | - | 4 - 5 | 711 | 85 | |
BC086992.1 | - | 4 - 5 | 630 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
