Assay Details
Gene Symbol: Tcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012670.1 | NP_036802.1 | 4 - 5 | 459 | 148 | |
GenBank mRNA | D90345.1 | - | 4 - 5 | 459 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
