Assay Details
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004078.1 | NP_001004078.1 | 8 - 9 | 1178 | 100 | |
GenBank mRNA | BC079441.1 | - | 8 - 9 | 1178 | 100 | |
FQ229253.1 | - | 3 - 4 | 397 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
