Assay Details
Gene Symbol: Fam118b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025283.1 | NP_001020454.1 | 4 - 5 | 712 | 159 | |
XM_006242784.2 | XP_006242846.1 | 5 - 6 | 792 | 159 | ||
XM_008766056.1 | XP_008764278.1 | 5 - 6 | 882 | 159 | ||
GenBank mRNA | BC098017.1 | - | 4 - 5 | 712 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
