Assay Details
Gene Symbol: Ntm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017354.1 | NP_059050.1 | 2 - 3 | 877 | 125 | |
XM_006242749.1 | XP_006242811.1 | 2 - 3 | 1195 | 125 | ||
XM_008766065.1 | XP_008764287.1 | 2 - 3 | 1195 | 125 | ||
GenBank mRNA | U16845.1 | - | 2 - 3 | 877 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
