Assay Details
Gene Symbol: Npr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012868.1 | NP_037000.1 | 7 - 8 | 1510 | 113 | |
XM_006232045.2 | XP_006232107.1 | 7 - 8 | 2097 | 113 | ||
GenBank mRNA | L27339.1 | - | 7 - 8 | 1510 | 113 | |
X75605.1 | - | 7 - 8 | 1531 | 113 | ||
X78595.1 | - | 1356 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
osteoclast proliferation
adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
negative regulation of adenylate cyclase activity
phospholipase C-activating G protein-coupled receptor signaling pathway
regulation of blood pressure
pancreatic juice secretion
regulation of osteoblast proliferation
positive regulation of urine volume
phosphatidylinositol-mediated signaling
negative regulation of smooth muscle cell proliferation
positive regulation of nitric-oxide synthase activity
G protein-coupled peptide receptor activity
natriuretic peptide receptor activity
peptide hormone binding
hormone binding
protein homodimerization activity
skeletal system development
osteoclast proliferation
adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
negative regulation of adenylate cyclase activity
phospholipase C-activating G protein-coupled receptor signaling pathway
regulation of blood pressure
pancreatic juice secretion
regulation of osteoblast proliferation
positive regulation of urine volume
phosphatidylinositol-mediated signaling
negative regulation of smooth muscle cell proliferation
positive regulation of nitric-oxide synthase activity
G protein-coupled peptide receptor activity
natriuretic peptide receptor activity
peptide hormone binding
hormone binding
protein homodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P036616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65146 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036616 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036616 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P036616 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P552517 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48296 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102348 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67496 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95518 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63886 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10821998 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821994 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821999 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821996 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821995 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821997 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821992 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821993 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10821991 | - | - |
Rat Genome 230 2.0 Array | 1396249_at | - | - |
Rat Genome 230 2.0 Array | 1369273_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1396249_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369273_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369273_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369273_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200021613.rn.1 | TC0200003085.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200021610.rn.1 | TC0200003085.rn.1 | RTA TC |
Rat Expression Set 230 | 1369273_a_at | - | - |
Rat Expression Set 230 | 1369273_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821991 | - | - |
Rat Genome U34 Set | rc_AA945907_at | - | - |
Rat Genome U34 Set | S79624_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20818260 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349272 | - | - |
RatRef-12 Expression BeadChip | ILMN_54504 | - | - |
