Assay Details
Gene Symbol: Amacr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012816.2 | NP_036948.2 | 2 - 3 | 397 | 63 | |
GenBank mRNA | AY325250.1 | - | 430 | 63 | ||
BC078885.1 | - | 2 - 3 | 365 | 63 | ||
BC091163.1 | - | 2 - 3 | 366 | 63 | ||
BC127505.1 | - | 2 - 3 | 386 | 63 | ||
FM061736.1 | - | 2 - 3 | 397 | 63 | ||
U89905.1 | - | 2 - 3 | 385 | 63 | ||
Y08172.1 | - | 2 - 3 | 375 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
