Assay Details
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165936.1 | NP_001159408.1 | 3 - 4 | 599 | 119 | |
NM_212532.2 | NP_997697.1 | 3 - 4 | 646 | 119 | ||
XM_008772714.1 | XP_008770936.1 | 4 - 5 | 745 | 119 | ||
XM_008772715.1 | XP_008770937.1 | 3 - 4 | 908 | 119 | ||
GenBank mRNA | BC086443.1 | - | 3 - 4 | 547 | 119 | |
BC097930.1 | - | 3 - 4 | 636 | 119 | ||
EV770973.1 | - | 3 - 4 | 599 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
