Assay Details
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004070.2 | NP_001004070.1 | 5 - 6 | 594 | 141 | |
NM_001165880.1 | NP_001159352.1 | 6 - 7 | 742 | 141 | ||
XM_006255977.2 | XP_006256039.1 | 6 - 7 | 1463 | 141 | ||
XM_006255978.2 | XP_006256040.1 | 7 - 8 | 1349 | 141 | ||
XM_006255979.2 | XP_006256041.1 | 7 - 8 | 845 | 141 | ||
GenBank mRNA | BC127463.1 | - | 6 - 7 | 742 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
