Assay Details
Gene Symbol: Tnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012675.3 | NP_036807.1 | 2 - 3 | 384 | 95 | |
GenBank mRNA | AF269159.1 | - | 2 - 3 | 240 | 95 | |
AF269160.1 | - | 2 - 3 | 240 | 95 | ||
AJ002278.1 | - | 2 - 3 | 240 | 95 | ||
BC107671.1 | - | 2 - 3 | 384 | 95 | ||
X66539.1 | - | 2 - 3 | 240 | 95 |
Gene Symbol: LOC103694380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772775.1 | XP_008770997.1 | 2 - 3 | 395 | 95 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
