Assay Details
Gene Symbol: Dxo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212497.2 | NP_997662.1 | 4 - 5 | 938 | 146 | |
GenBank mRNA | BC082083.1 | - | 4 - 5 | 938 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
