Assay Details
Gene Symbol: Stk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013197.2 | NP_001013215.2 | 1 - 2 | 149 | 153 | |
GenBank mRNA | BC061571.1 | - | 1 - 2 | 153 | 153 | |
CO398403.1 | - | 1 - 2 | 142 | 153 |
Gene Symbol: LOC103694374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772703.1 | XP_008770925.1 | 153 | 153 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC061571.1, NM_001013197.2, XM_008772703.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
