Assay Details
Gene Symbol: Cfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212466.3 | NP_997631.2 | 11 - 12 | 1710 | 63 | |
GenBank mRNA | BC087089.1 | - | 11 - 12 | 1710 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
