Assay Details
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109137.1 | NP_001102607.1 | 2 - 3 | 388 | 82 | |
XM_006227887.2 | XP_006227949.1 | 3 - 4 | 2741 | 82 | ||
XM_008758736.1 | XP_008756958.1 | 3 - 4 | 2225 | 82 | ||
XM_008758737.1 | XP_008756959.1 | 3 - 4 | 2913 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
