Assay Details
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019357.1 | NP_062230.1 | 11 - 12 | 1821 | 90 | |
GenBank mRNA | AY428869.1 | - | 11 - 12 | 1406 | 90 | |
BC081958.1 | - | 11 - 12 | 1821 | 90 | ||
X67788.1 | - | 1 - 2 | 62 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
