Assay Details
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019357.1 | NP_062230.1 | 8 - 9 | 1445 | 67 | |
GenBank mRNA | AF450298.1 | - | 8 - 9 | 1044 | 67 | |
AY428869.1 | - | 8 - 9 | 1030 | 67 | ||
BC081958.1 | - | 8 - 9 | 1445 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
