Assay Details
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254965.2 | XP_006255027.1 | 1 - 2 | 72 | 55 | |
GenBank mRNA | BC078941.1 | - | 1 - 2 | 46 | 55 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC078941.1, XM_006254965.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
