Assay Details
Gene Symbol: Slc39a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108728.1 | NP_001102198.1 | 7 - 8 | 1403 | 65 | |
XM_006240760.2 | XP_006240822.1 | 8 - 9 | 1308 | 65 | ||
XM_006240761.2 | XP_006240823.1 | 8 - 9 | 1320 | 65 | ||
XM_006240762.2 | XP_006240824.1 | 9 - 10 | 1428 | 65 | ||
XM_006240763.2 | XP_006240825.1 | 9 - 10 | 1416 | 65 | ||
XM_006240764.2 | XP_006240826.1 | 7 - 8 | 1212 | 65 | ||
XM_006240766.2 | XP_006240828.1 | 8 - 9 | 1523 | 65 | ||
XM_006240767.2 | XP_006240829.1 | 6 - 7 | 972 | 65 | ||
XM_008765027.1 | XP_008763249.1 | 8 - 9 | 1342 | 65 | ||
XM_008765028.1 | XP_008763250.1 | 8 - 9 | 1308 | 65 | ||
GenBank mRNA | FQ220010.1 | - | 4 - 5 | 498 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
