Assay Details
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105939.2 | NP_001099409.1 | 2 - 3 | 179 | 95 | |
NM_001198562.1 | NP_001185491.1 | 2 - 3 | 141 | 95 | ||
GenBank mRNA | BC166934.1 | - | 2 - 3 | 129 | 95 | |
BP494723.1 | - | 2 - 3 | 163 | 95 | ||
FQ221678.1 | - | 2 - 3 | 174 | 95 | ||
FQ225664.1 | - | 2 - 3 | 115 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
