Assay Details
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105939.2 | NP_001099409.1 | 7 - 8 | 731 | 141 | |
NM_001198562.1 | NP_001185491.1 | 7 - 8 | 693 | 141 | ||
NM_001198580.1 | NP_001185509.1 | 7 - 8 | 6471 | 141 | ||
GenBank mRNA | BC166934.1 | - | 7 - 8 | 681 | 141 | |
FQ215294.1 | - | 6 - 7 | 1212 | 141 | ||
FQ221678.1 | - | 7 - 8 | 726 | 141 | ||
FQ231879.1 | - | 7 - 8 | 720 | 141 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
