Assay Details
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105939.2 | NP_001099409.1 | 1 - 2 | 109 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105939.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
