Assay Details
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033070.1 | NP_001028242.1 | 2 - 3 | 295 | 126 | |
XM_006240781.2 | XP_006240843.1 | 2 - 3 | 324 | 126 | ||
XM_008765031.1 | XP_008763253.1 | 2 - 3 | 319 | 126 | ||
GenBank mRNA | BC100070.1 | - | 2 - 3 | 295 | 126 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
