Assay Details
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033070.1 | NP_001028242.1 | 8 - 9 | 588 | 102 | |
XM_008765031.1 | XP_008763253.1 | 8 - 9 | 612 | 102 | ||
GenBank mRNA | BC100070.1 | - | 8 - 9 | 588 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
