Assay Details
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033070.1 | NP_001028242.1 | 1 - 2 | 198 | 89 | |
XM_006240781.2 | XP_006240843.1 | 1 - 2 | 227 | 89 | ||
XM_008765031.1 | XP_008763253.1 | 1 - 2 | 222 | 89 | ||
GenBank mRNA | BC100070.1 | - | 1 - 2 | 198 | 89 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC100070.1, NM_001033070.1, XM_006240781.2, XM_008765031.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
