Assay Details
Gene Symbol: Slc22a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019230.1 | NP_062103.1 | 2 - 3 | 907 | 95 | |
GenBank mRNA | AF055286.1 | - | 2 - 3 | 907 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
