Assay Details
Gene Symbol: Pacrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077677.1 | NP_001071145.1 | 650 | 104 | ||
GenBank mRNA | BC127534.1 | - | 650 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
