Assay Details
Gene Symbol: Gtf2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007711.1 | NP_001007712.1 | 10 - 11 | 1223 | 61 | |
XM_006244321.2 | XP_006244383.1 | 11 - 12 | 1346 | 61 | ||
GenBank mRNA | BC079136.1 | - | 10 - 11 | 1223 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
