Assay Details
Gene Symbol: Sh3gl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031239.2 | NP_112518.1 | 7 - 8 | 862 | 71 | |
GenBank mRNA | AB008161.1 | - | 7 - 8 | 862 | 71 | |
AF009602.1 | - | 7 - 8 | 847 | 71 | ||
BC070893.1 | - | 7 - 8 | 858 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
