Assay Details
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004203.3 | NP_001004203.3 | 5 - 6 | 784 | 93 | |
XM_006249005.2 | XP_006249067.1 | 5 - 6 | 906 | 93 | ||
XM_006249006.2 | XP_006249068.1 | 5 - 6 | 789 | 93 | ||
GenBank mRNA | BC078973.1 | - | 5 - 6 | 784 | 93 | |
DQ278199.1 | - | 5 - 6 | 832 | 93 | ||
DQ278200.1 | - | 1377 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
