Assay Details
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044225.1 | NP_001037690.1 | 12 - 13 | 1383 | 104 | |
XM_008769061.1 | XP_008767283.1 | 12 - 13 | 1423 | 104 | ||
XM_008769062.1 | XP_008767284.1 | 12 - 13 | 1422 | 104 | ||
GenBank mRNA | BC100108.1 | - | 12 - 13 | 1383 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
