Assay Details
Gene Symbol: Vamp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053531.1 | NP_445983.1 | 4 - 5 | 433 | 163 | |
XM_006249016.2 | XP_006249078.1 | 5 - 6 | 586 | 163 | ||
GenBank mRNA | AF281632.1 | - | 4 - 5 | 433 | 163 | |
FQ228923.1 | - | 5 - 6 | 546 | 163 | ||
FQ230649.1 | - | 4 - 5 | 357 | 163 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
