Assay Details
Gene Symbol: Ftsj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256749.2 | XP_006256811.1 | 3 - 4 | 646 | 115 | |
XM_006256750.2 | XP_006256812.1 | 3 - 4 | 646 | 115 | ||
XM_006256753.2 | XP_006256815.1 | 3 - 4 | 494 | 115 | ||
XM_008773086.1 | XP_008771308.1 | 3 - 4 | 867 | 115 | ||
XM_008773087.1 | XP_008771309.1 | 3 - 4 | 643 | 115 | ||
XM_008773088.1 | XP_008771310.1 | 3 - 4 | 270 | 115 | ||
XM_008773090.1 | XP_008771312.1 | 3 - 4 | 493 | 115 | ||
XM_008773091.1 | XP_008771313.1 | 3 - 4 | 652 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
