Assay Details
Gene Symbol: Ftsj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256750.2 | XP_006256812.1 | 11 - 12 | 1407 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
