Assay Details
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133538.2 | NP_598222.2 | 11 - 12 | 1460 | 107 | |
XM_006257427.2 | XP_006257489.1 | 10 - 11 | 1328 | 107 | ||
XM_006257428.2 | XP_006257490.1 | 10 - 11 | 1446 | 107 | ||
XM_006257430.2 | XP_006257492.1 | 9 - 10 | 1259 | 107 | ||
XM_006257431.2 | XP_006257493.1 | 7 - 8 | 967 | 107 | ||
XM_008773388.1 | XP_008771610.1 | 10 - 11 | 1600 | 107 | ||
XM_008773389.1 | XP_008771611.1 | 11 - 12 | 1487 | 107 | ||
XM_008773390.1 | XP_008771612.1 | 7 - 8 | 1061 | 107 | ||
XM_008773391.1 | XP_008771613.1 | 7 - 8 | 1668 | 107 | ||
GenBank mRNA | AF448818.1 | - | 11 - 12 | 1460 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
