Assay Details
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012762.2 | NP_036894.2 | 1 - 2 | 276 | 127 | |
GenBank mRNA | D85899.1 | - | 1 - 2 | 276 | 127 | |
U14647.1 | - | 1 - 2 | 276 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
