Assay Details
Gene Symbol: Aasdhppt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106798.2 | NP_001100268.2 | 2 - 3 | 429 | 126 | |
GenBank mRNA | BC098729.1 | - | 1 - 2 | 785 | 126 | |
BC166800.1 | - | 2 - 3 | 429 | 126 | ||
FQ209549.1 | - | 2 - 3 | 445 | 126 | ||
FQ214274.1 | - | 2 - 3 | 442 | 126 | ||
FQ214451.1 | - | 2 - 3 | 443 | 126 | ||
FQ220704.1 | - | 2 - 3 | 443 | 126 | ||
FQ230080.1 | - | 2 - 3 | 448 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
