Assay Details
Gene Symbol: Gucy1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023956.1 | NP_076446.1 | 5 - 6 | 2106 | 108 | |
GenBank mRNA | AB096080.1 | - | 5 - 6 | 2106 | 108 | |
AB097860.1 | - | 5 - 6 | 1686 | 108 | ||
AF109963.2 | - | 5 - 6 | 2106 | 108 | ||
AJ001041.1 | - | 1 - 2 | 92 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
