Assay Details
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108989.1 | NP_001102459.1 | 6 - 7 | 1770 | 91 | |
GenBank mRNA | BC166847.1 | - | 6 - 7 | 1753 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
