Assay Details
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153296.2 | NP_695208.2 | 3 - 4 | 367 | 143 | |
XM_006235675.2 | XP_006235737.1 | 4 - 5 | 446 | 143 | ||
XM_006235676.2 | XP_006235738.1 | 4 - 5 | 463 | 143 | ||
GenBank mRNA | AF537333.1 | - | 3 - 4 | 355 | 143 | |
BC104676.1 | - | 3 - 4 | 367 | 143 | ||
FQ227101.1 | - | 4 - 5 | 453 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
