Assay Details
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235675.2 | XP_006235737.1 | 1 - 2 | 84 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235675.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
