Assay Details
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017125.3 | NP_058821.1 | 5 - 6 | 488 | 82 | |
XM_008765024.1 | XP_008763246.1 | 5 - 6 | 550 | 82 | ||
XM_008765025.1 | XP_008763247.1 | 5 - 6 | 753 | 82 | ||
GenBank mRNA | AF159104.1 | - | 1 - 2 | 112 | 82 | |
BC063173.1 | - | 5 - 6 | 491 | 82 | ||
FQ214468.1 | - | 5 - 6 | 555 | 82 | ||
FQ221333.1 | - | 5 - 6 | 492 | 82 | ||
FQ222038.1 | - | 5 - 6 | 471 | 82 | ||
FQ230116.1 | - | 5 - 6 | 459 | 82 | ||
X61654.1 | - | 5 - 6 | 488 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
